Assay Details
| Gene Symbol: Nudt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025539.2 | NP_079815.2 | 1 - 2 | 34 | 69 | |
| GenBank mRNA | AK009933.1 | - | 1 - 2 | 32 | 69 | |
| AK018771.1 | - | 1 - 2 | 32 | 69 | ||
| AK132532.1 | - | 35 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009933.1, AK018771.1, AK132532.1, NM_025539.2 transcripts.
Set Membership: |
|

Nudt2
Genomic Map