Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900009311.mm.1 |
NM_177909.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009311.mm.1 |
NM_177909.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009311.mm.1 |
TC0900001182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009309.mm.1 |
NM_177909.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009309.mm.1 |
NM_177909.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009309.mm.1 |
TC0900001182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009312.mm.1 |
NM_177909.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009312.mm.1 |
NM_177909.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009312.mm.1 |
TC0900001182.mm.1 |
MTA TC |