Assay Details
| Gene Symbol: Prss40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009356.2 | NP_033382.2 | 1 - 2 | 213 | 77 | |
| GenBank mRNA | BC145989.1 | - | 1 - 2 | 228 | 40131292 | 77 |
More Information
Set Membership: |
|

Prss40
Genomic Map