Assay Details
| Gene Symbol: Prss40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009356.2 | NP_033382.2 | 4 - 5 | 822 | 79 | |
| GenBank mRNA | AB008911.1 | - | 4 - 5 | 805 | 79 | |
| BC145989.1 | - | 4 - 5 | 837 | 40131292 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059030 | AK132909.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556639 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352014 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106148 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2059030 | AK132909.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P197805 | AB008911.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139444 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106148 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04059418 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04059441 | - | - |
| Whole Mouse Genome G4122A | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48480 | AK006524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48480 | NM_009356.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48480 | BC145989.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P197805 | AB008911.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117084 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102462 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106148 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00546962 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00546961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00219332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00546963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00769455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00299646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00546963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00804932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00301013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00219332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00769449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00546961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00769448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00718407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00566914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10353790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353788 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432486_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353786 | - | - |
| Mouse Genome 430 2.0 Array | 1432486_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432486_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21756_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21756_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100017269.mm.1 | NM_009356.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017269.mm.1 | NM_009356.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017269.mm.1 | TC0100002154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100017270.mm.1 | NM_009356.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017270.mm.1 | NM_009356.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017270.mm.1 | TC0100002154.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432486_PM_a_at | - | - |
| Mouse Expression Set 430 | 1432486_a_at | - | - |
| Murine Genome U74v2 Set | 94741_at | - | - |
| Murine Genome U74v2 Set | 94741_PM_at | - | - |
| Murine Genome U74v2 Set | 167738_f_at | - | - |
| Murine Genome U74v2 Set | 167738_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432486_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432486_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432486_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432486_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 509267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2340403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161847 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771861 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771861 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161847 | - | - |
| mouse-6 v1.1 (Illumina) | scl16882.5.1_130-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6678294-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16882.5.1_130-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2771861 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771861 | - | - |

Prss40
Genomic Map