Assay Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011206.2 | NP_035336.2 | 12 - 13 | 1092 | 100 | |
| GenBank mRNA | AK144353.1 | - | 12 - 13 | 1074 | 100 | |
| BC008512.1 | - | 12 - 13 | 1062 | 3591418 | 100 | |
| CD740968.1 | - | 3 - 4 | 100 | |||
| U35124.1 | - | 12 - 13 | 1066 | 100 | ||
| U49853.1 | - | 12 - 13 | 1032 | 100 | ||
| U52523.1 | - | 12 - 13 | 1080 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn18
Genomic Map