Assay Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011206.2 | NP_035336.2 | 3 - 4 | 352 | 63 | |
| GenBank mRNA | AK144353.1 | - | 3 - 4 | 334 | 63 | |
| U35124.1 | - | 3 - 4 | 326 | 63 | ||
| U49853.1 | - | 3 - 4 | 292 | 63 | ||
| U52523.1 | - | 3 - 4 | 340 | 63 | ||
More Information
Set Membership: |
|

Ptpn18
Genomic Map