Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100000801.mm.1 |
NM_031402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000801.mm.1 |
NM_031402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000801.mm.1 |
TC0100000113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000802.mm.1 |
NM_031402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000802.mm.1 |
NM_031402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000802.mm.1 |
TC0100000113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000799.mm.1 |
TC0100000113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000800.mm.1 |
NM_031402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000800.mm.1 |
NM_031402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000800.mm.1 |
TC0100000113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000798.mm.1 |
TC0100000113.mm.1 |
MTA TC |