Assay Details
| Gene Symbol: Stard8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199018.2 | NP_950183.1 | 2 - 3 | 290 | 70 | |
| GenBank mRNA | AK133927.1 | - | 2 - 3 | 238 | 70 | |
| AK149830.1 | - | 2 - 3 | 221 | 70 | ||
| AK150259.1 | - | 2 - 3 | 221 | 70 | ||
| BC078461.1 | - | 1 - 2 | 55 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133927.1, AK149830.1, AK150259.1, NM_199018.2 transcripts.
Set Membership: |
|

Stard8
Genomic Map