Assay Details
| Gene Symbol: Stard8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199018.2 | NP_950183.1 | 5 - 6 | 1842 | 87 | |
| GenBank mRNA | AK133927.1 | - | 5 - 6 | 1790 | 87 | |
| AK149830.1 | - | 5 - 6 | 1773 | 87 | ||
| AK150259.1 | - | 5 - 6 | 1773 | 87 | ||
| BC034186.1 | - | 1 - 2 | 964 | 87 | ||
| BC062944.1 | - | 5 - 6 | 1757 | 30357177 | 87 | |
| BC078461.1 | - | 4 - 5 | 1607 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard8
Genomic Map