Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008765.mm.1 |
NM_029852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008765.mm.1 |
NM_029852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008765.mm.1 |
TC1000001153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008766.mm.1 |
NM_029852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008766.mm.1 |
NM_029852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008766.mm.1 |
TC1000001153.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008764.mm.1 |
NM_029852.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008764.mm.1 |
NM_029852.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008764.mm.1 |
TC1000001153.mm.1 |
MTA TC |