Assay Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008777.3 | NP_032803.2 | 6 - 7 | 925 | 57 | |
| GenBank mRNA | AK145839.1 | - | 6 - 7 | 790 | 57 | |
| AK149519.1 | - | 6 - 7 | 791 | 57 | ||
| BC013458.1 | - | 6 - 7 | 783 | 4223155 | 57 | |
| X51942.1 | - | 6 - 7 | 750 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pah
Genomic Map