Assay Details
| Gene Symbol: Tceb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011958.1 | - | 1 - 2 | 54 | 124 | |
| AK013089.1 | - | 1 - 2 | 51 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011958.1,AK013089.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tceb1
Genomic Map