Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 22 - 23 | 3278 | 141 | |
| GenBank mRNA | AK003230.1 | - | 6 - 7 | 693 | 141 | |
| AK081350.1 | - | 22 - 23 | 3470 | 142 | ||
| AK090116.1 | - | 22 - 23 | 3155 | 141 | ||
| AK154106.1 | - | 22 - 23 | 3263 | 141 | ||
| AK165230.1 | - | 738 | 141 | |||
| BC127073.1 | - | 21 - 22 | 2927 | 40110218 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map