Assay Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028315.2 | NP_082591.2 | 12 - 13 | 1766 | 136 | |
| GenBank mRNA | AK032091.1 | - | 7 - 8 | 856 | 136 | |
| AK038313.1 | - | 12 - 13 | 1636 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dis3
Genomic Map