Assay Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028315.2 | NP_082591.2 | 15 - 16 | 2074 | 121 | |
| GenBank mRNA | AK032091.1 | - | 10 - 11 | 1164 | 121 | |
| AK038313.1 | - | 15 - 16 | 1944 | 120 | ||
| BC027357.1 | - | 14 - 15 | 1959 | 4952415 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3
Genomic Map