Assay Details
Gene Symbol: Irg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008392.1 | NP_032418.1 | 2 - 3 | 221 | 66 | |
GenBank mRNA | AK036446.1 | - | 2 - 3 | 257 | 66 | |
AK152177.1 | - | 2 - 3 | 220 | 66 | ||
AK152248.1 | - | 2 - 3 | 220 | 66 | ||
AK152332.1 | - | 2 - 3 | 220 | 66 | ||
AK152635.1 | - | 2 - 3 | 220 | 66 | ||
AK153013.1 | - | 2 - 3 | 220 | 66 | ||
AK172001.1 | - | 2 - 3 | 220 | 66 | ||
AK172298.1 | - | 2 - 3 | 220 | 66 | ||
L38281.1 | - | 2 - 3 | 438 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
