Assay Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145465.2 | NP_663440.1 | 4 - 5 | 732 | 142 | |
| GenBank mRNA | AK154596.1 | - | 4 - 5 | 837 | 142 | |
| AK154959.1 | - | 4 - 5 | 733 | 142 | ||
| AY188357.1 | - | 4 - 5 | 742 | 142 | ||
| BC004650.1 | - | 4 - 5 | 564 | 3482980 | 142 | |
More Information
Set Membership: |
|

Stk24
Genomic Map