Assay Details
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145465.2 | NP_663440.1 | 8 - 9 | 1343 | 97 | |
GenBank mRNA | AK154596.1 | - | 8 - 9 | 1448 | 97 | |
AK154959.1 | - | 8 - 9 | 1344 | 97 | ||
AY188357.1 | - | 8 - 9 | 1353 | 97 | ||
BC004650.1 | - | 8 - 9 | 1175 | 3482980 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
