Assay Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145465.2 | NP_663440.1 | 9 - 10 | 1411 | 74 | |
| GenBank mRNA | AK154596.1 | - | 9 - 10 | 1516 | 74 | |
| AK154959.1 | - | 9 - 10 | 1412 | 74 | ||
| AY188357.1 | - | 9 - 10 | 1421 | 74 | ||
| BC004650.1 | - | 9 - 10 | 1243 | 3482980 | 74 | |
More Information
Set Membership: |
|

Stk24
Genomic Map