Assay Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033314.3 | NP_201571.2 | 3 - 4 | 526 | 93 | |
| GenBank mRNA | AF323958.1 | - | 3 - 4 | 395 | 93 | |
| AK166724.1 | - | 3 - 4 | 527 | 93 | ||
| AK169706.1 | - | 3 - 4 | 529 | 93 | ||
| BC035200.1 | - | 516 | 3158233 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco2a1
Genomic Map