Assay Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033314.3 | NP_201571.2 | 7 - 8 | 1069 | 84 | |
| GenBank mRNA | AF323958.1 | - | 7 - 8 | 938 | 84 | |
| AK087350.1 | - | 7 - 8 | 1075 | 84 | ||
| AK166724.1 | - | 7 - 8 | 1070 | 84 | ||
| AK169706.1 | - | 7 - 8 | 1072 | 84 | ||
| BC035200.1 | - | 1059 | 3158233 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco2a1
Genomic Map