Assay Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033314.3 | NP_201571.2 | 8 - 9 | 1236 | 99 | |
| GenBank mRNA | AF323958.1 | - | 8 - 9 | 1105 | 99 | |
| AK166724.1 | - | 8 - 9 | 1237 | 99 | ||
| AK169706.1 | - | 8 - 9 | 1239 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco2a1
Genomic Map