Assay Details
| Gene Symbol: Hdac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027382.3 | NP_081658.1 | 7 - 8 | 820 | 107 | |
| GenBank mRNA | AK011332.1 | - | 7 - 8 | 821 | 107 | |
| AK131998.1 | - | 7 - 8 | 1033 | 107 | ||
| BC061257.1 | - | 7 - 8 | 792 | 30251620 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac8
Genomic Map