Assay Details
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177352.4 | NP_796326.1 | 12 - 13 | 1181 | 83 | |
GenBank mRNA | AK049275.1 | - | 12 - 13 | 1182 | 83 | |
AK076293.1 | - | 12 - 13 | 1210 | 83 | ||
AK160477.1 | - | 12 - 13 | 1236 | 83 | ||
AK162119.1 | - | 12 - 13 | 1182 | 83 | ||
AK167217.1 | - | 12 - 13 | 1201 | 83 | ||
BC050033.1 | - | 3 - 4 | 248 | 83 | ||
BC116302.1 | - | 12 - 13 | 1178 | 40095102 | 83 | |
BC116303.1 | - | 11 - 12 | 1084 | 40095106 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
