Assay Details
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177352.4 | NP_796326.1 | 14 - 15 | 1341 | 65 | |
GenBank mRNA | AK049275.1 | - | 14 - 15 | 1342 | 65 | |
AK076293.1 | - | 14 - 15 | 1370 | 65 | ||
AK160477.1 | - | 14 - 15 | 1396 | 65 | ||
AK162119.1 | - | 14 - 15 | 1342 | 65 | ||
AK163031.1 | - | 14 - 15 | 66 | |||
AK167217.1 | - | 14 - 15 | 1361 | 65 | ||
BC050033.1 | - | 5 - 6 | 408 | 65 | ||
BC116302.1 | - | 14 - 15 | 1338 | 40095102 | 65 | |
BC116303.1 | - | 13 - 14 | 1244 | 40095106 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
