Assay Details
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008448.3 | NP_032474.2 | 10 - 11 | 1380 | 82 | |
GenBank mRNA | AK085221.1 | - | 2 - 3 | 206 | 82 | |
AK133535.1 | - | 10 - 11 | 1377 | 82 | ||
AK136307.1 | - | 10 - 11 | 1377 | 82 | ||
BC013248.1 | - | 10 - 11 | 1336 | 82 | ||
BC013817.1 | - | 10 - 11 | 1346 | 82 | ||
BC025864.1 | - | 10 - 11 | 1349 | 82 | ||
BC040800.1 | - | 10 - 11 | 1370 | 82 | ||
BC069920.1 | - | 10 - 11 | 1356 | 82 | ||
BC090841.1 | - | 10 - 11 | 1356 | 30543821 | 82 | |
L27153.1 | - | 10 - 11 | 974 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
