Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 2 - 3 | 638 | 94 | |
| GenBank mRNA | AK133535.1 | - | 2 - 3 | 635 | 94 | |
| AK136307.1 | - | 2 - 3 | 635 | 94 | ||
| AK185672.1 | - | 1 - 2 | 94 | |||
| BC013248.1 | - | 2 - 3 | 594 | 94 | ||
| BC013817.1 | - | 2 - 3 | 604 | 94 | ||
| BC025864.1 | - | 2 - 3 | 607 | 94 | ||
| BC040800.1 | - | 2 - 3 | 628 | 94 | ||
| BC069920.1 | - | 2 - 3 | 614 | 94 | ||
| BC090841.1 | - | 2 - 3 | 614 | 30543821 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif5b
Genomic Map