Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 8 - 9 | 1123 | 80 | |
| GenBank mRNA | AK133535.1 | - | 8 - 9 | 1120 | 80 | |
| AK136307.1 | - | 8 - 9 | 1120 | 80 | ||
| BC013248.1 | - | 8 - 9 | 1079 | 80 | ||
| BC013817.1 | - | 8 - 9 | 1089 | 80 | ||
| BC025864.1 | - | 8 - 9 | 1092 | 80 | ||
| BC040800.1 | - | 8 - 9 | 1113 | 80 | ||
| BC069920.1 | - | 8 - 9 | 1099 | 80 | ||
| BC090841.1 | - | 8 - 9 | 1099 | 30543821 | 80 | |
| L27153.1 | - | 8 - 9 | 717 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif5b
Genomic Map