Assay Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122960.1 | NP_001116432.1 | 5 - 6 | 576 | 90 | |
| NM_026070.3 | NP_080346.1 | 5 - 6 | 579 | 90 | ||
| GenBank mRNA | AK004206.1 | - | 5 - 6 | 494 | 90 | |
| AK009358.1 | - | 5 - 6 | 475 | 90 | ||
| AK013835.1 | - | 5 - 6 | 584 | 90 | ||
| AK158822.1 | - | 5 - 6 | 580 | 90 | ||
| CK795584.1 | - | 5 - 6 | 528 | 90 | ||
More Information
Set Membership: |
|

Ccdc53
Genomic Map