Assay Details
| Gene Symbol: Slc6a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175328.3 | NP_780537.1 | 2 - 3 | 440 | 134 | |
| GenBank mRNA | AY149280.1 | - | 2 - 3 | 405 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY149280.1, NM_175328.3 transcripts.
Set Membership: |
|

Slc6a15
Genomic Map