| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000009337.mm.1 |
NM_001252330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009337.mm.1 |
NM_001252330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009337.mm.1 |
NM_175328.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009337.mm.1 |
NM_175328.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009337.mm.1 |
TC1000001244.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000009338.mm.1 |
NM_001252330.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009338.mm.1 |
NM_001252330.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009338.mm.1 |
NM_175328.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009338.mm.1 |
NM_175328.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000009338.mm.1 |
TC1000001244.mm.1 |
MTA TC |