Assay Details
| Gene Symbol: Atl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286647.1 | NP_001273576.1 | 3 - 4 | 451 | 125 | |
| NM_019717.3 | NP_062691.3 | 4 - 5 | 696 | 125 | ||
| NM_178050.4 | NP_835151.2 | 3 - 4 | 451 | 125 | ||
| GenBank mRNA | AK038697.1 | - | 3 - 4 | 379 | 125 | |
| AK135988.1 | - | 3 - 4 | 342 | 125 | ||
| BC060501.1 | - | 4 - 5 | 601 | 5362692 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135988.1 transcripts.
Set Membership: |
|

Atl2
Genomic Map