Assay Details
Gene Symbol: Ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC116213.1 | - | 2 - 3 | 214 | 81 | |
BC116214.1 | - | 2 - 3 | 214 | 81 | ||
BC128473.1 | - | 2 - 3 | 214 | 81 | ||
BC128474.1 | - | 2 - 3 | 214 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
