Assay Details
Gene Symbol: Qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027455.2 | NP_081731.1 | 5 - 6 | 1186 | 150 | |
GenBank mRNA | AK017598.1 | - | 5 - 6 | 849 | 150 | |
AK037197.1 | - | 5 - 6 | 839 | 150 | ||
AK045974.1 | - | 5 - 6 | 1187 | 150 | ||
AK148394.1 | - | 5 - 6 | 857 | 150 | ||
BC020023.1 | - | 4 - 5 | 757 | 3491756 | 150 | |
BC151027.1 | - | 5 - 6 | 930 | 9087940 | 150 | |
BC151029.1 | - | 5 - 6 | 930 | 9087942 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
