Assay Details
| Gene Symbol: Tfap2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153154.2 | NP_694794.1 | 4 - 5 | 991 | 67 | |
| GenBank mRNA | AF421891.1 | - | 4 - 5 | 1018 | 67 | |
| AK132112.1 | - | 4 - 5 | 993 | 67 | ||
| BC152324.1 | - | 4 - 5 | 940 | 40141049 | 67 | |
| BC152325.1 | - | 4 - 5 | 940 | 40141051 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfap2d
Genomic Map