Assay Details
Gene Symbol: Sp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166385.2 | NP_001159857.2 | 1 - 2 | 212 | 64 | |
NM_009239.4 | NP_033265.4 | 1 - 2 | 212 | 64 | ||
GenBank mRNA | BC076630.1 | - | 1 - 2 | 142 | 30637762 | 64 |
U62522.1 | - | 1 - 2 | 141 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC076630.1, NM_001166385.2, NM_009239.4, U62522.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
