Assay Details
Gene Symbol: Abcb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029961.2 | NP_084237.1 | 9 - 10 | 1059 | 86 | |
GenBank mRNA | AY766239.1 | - | 9 - 10 | 1023 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
