Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100016685.mm.1 |
NM_001160139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016685.mm.1 |
NM_001160139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016685.mm.1 |
NM_023872.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016685.mm.1 |
NM_023872.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016685.mm.1 |
TC0100002042.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016683.mm.1 |
NM_001160139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016683.mm.1 |
NM_001160139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016683.mm.1 |
TC0100002042.mm.1 |
MTA TC |