Assay Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177278.5 | NP_796252.2 | 7 - 8 | 1041 | 77 | |
| GenBank mRNA | AK053068.1 | - | 7 - 8 | 1042 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl4
Genomic Map