Assay Details
Gene Symbol: Abcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033336.3 | NP_001028508.2 | 24 - 25 | 3129 | 63 | |
NM_001163675.1 | NP_001157147.1 | 24 - 25 | 3111 | 63 | ||
NM_001163676.1 | NP_001157148.1 | 23 - 24 | 2904 | 63 | ||
GenBank mRNA | AK032802.1 | - | 6 - 7 | 692 | 63 | |
AK052778.1 | - | 24 - 25 | 3113 | 63 | ||
AK157723.1 | - | 24 - 25 | 3116 | 63 | ||
BC150822.1 | - | 24 - 25 | 3083 | 9087733 | 63 | |
BC171974.1 | - | 23 - 24 | 2858 | 9054260 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
