Assay Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252478.1 | NP_001239407.1 | 1 - 2 | 62 | 107 | |
| NM_144798.6 | NP_659047.2 | 1 - 2 | 62 | 107 | ||
| GenBank mRNA | AK082907.1 | - | 1 - 2 | 39 | 107 | |
| AK150342.1 | - | 1 - 2 | 38 | 107 | ||
| AK151120.1 | - | 1 - 2 | 38 | 107 | ||
| AK160903.1 | - | 1 - 2 | 50 | 108 | ||
| BC068169.1 | - | 1 - 2 | 51 | 30542277 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082907.1, AK150342.1, AK151120.1, BC068169.1, NM_001252478.1, NM_144798.6 transcripts.
Set Membership: |
|

Slc30a6
Genomic Map