Assay Details
| Gene Symbol: Twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011876.3 | NP_036006.1 | 4 - 5 | 533 | 81 | |
| GenBank mRNA | AK002699.1 | - | 3 - 4 | 448 | 81 | |
| AK003997.1 | - | 3 - 4 | 428 | 81 | ||
| AK084026.1 | - | 4 - 5 | 533 | 81 | ||
| AK087422.1 | - | 3 - 4 | 448 | 81 | ||
| AK155204.1 | - | 4 - 5 | 516 | 81 | ||
| AK157886.1 | - | 4 - 5 | 478 | 81 | ||
| AK166900.1 | - | 4 - 5 | 501 | 81 | ||
| AK170441.1 | - | 4 - 5 | 481 | 81 | ||
| AY267189.1 | - | 4 - 5 | 381 | 81 | ||
| BC003338.1 | - | 4 - 5 | 480 | 3257230 | 81 | |
| Y17808.1 | - | 4 - 5 | 398 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Twf2
Genomic Map