Assay Details
| Gene Symbol: Slco5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172841.2 | NP_766429.2 | 4 - 5 | 2072 | 72 | |
| GenBank mRNA | AK041736.1 | - | 4 - 5 | 2073 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco5a1
Genomic Map