Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700011379.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011379.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011379.mm.1 |
TC1700001215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011378.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011378.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011378.mm.1 |
TC1700001215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011382.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011382.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011382.mm.1 |
TC1700001215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011383.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011383.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011383.mm.1 |
TC1700001215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011377.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011377.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011377.mm.1 |
TC1700001215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011380.mm.1 |
NM_001145452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011380.mm.1 |
NM_001145452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011380.mm.1 |
TC1700001215.mm.1 |
MTA TC |