Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 16 - 17 | 1733 | 69 | |
| GenBank mRNA | AK076357.1 | - | 17 - 18 | 1763 | 69 | |
| AK152868.1 | - | 16 - 17 | 1721 | 69 | ||
| BC049929.1 | - | 4 - 5 | 247 | 69 | ||
| BC079656.1 | - | 16 - 17 | 1733 | 6854039 | 69 | |
More Information
Set Membership: |
|

Ddx11
Genomic Map