Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 7 - 8 | 887 | 78 | |
| GenBank mRNA | AK049419.1 | - | 7 - 8 | 897 | 78 | |
| AK076357.1 | - | 7 - 8 | 916 | 78 | ||
| AK152868.1 | - | 7 - 8 | 874 | 78 | ||
| BC079656.1 | - | 7 - 8 | 887 | 6854039 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map