Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 9 - 10 | 1177 | 65 | |
| GenBank mRNA | AK152868.1 | - | 9 - 10 | 1164 | 65 | |
| BC079656.1 | - | 9 - 10 | 1177 | 6854039 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map