Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100002157.mm.1 |
NM_008362.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002157.mm.1 |
NM_008362.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002157.mm.1 |
TC0100000295.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002158.mm.1 |
NM_008362.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002158.mm.1 |
NM_008362.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002158.mm.1 |
TC0100000295.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002160.mm.1 |
NM_001123382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002160.mm.1 |
NM_001123382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002160.mm.1 |
NM_008362.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002160.mm.1 |
NM_008362.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002160.mm.1 |
TC0100000295.mm.1 |
MTA TC |