Assay Details
| Gene Symbol: Il1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123382.1 | NP_001116854.1 | 8 - 9 | 1287 | 104 | |
| NM_008362.2 | NP_032388.1 | 8 - 9 | 1218 | 104 | ||
| GenBank mRNA | AK048550.1 | - | 8 - 9 | 1205 | 104 | |
| AK054137.1 | - | 8 - 9 | 1288 | 104 | ||
| BC109135.2 | - | 8 - 9 | 1124 | 40039321 | 104 | |
| M20658.1 | - | 8 - 9 | 1218 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1r1
Genomic Map