Assay Details
| Gene Symbol: Il1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123382.1 | NP_001116854.1 | 9 - 10 | 1441 | 139 | |
| NM_008362.2 | NP_032388.1 | 9 - 10 | 1372 | 139 | ||
| GenBank mRNA | AK048550.1 | - | 9 - 10 | 1359 | 139 | |
| AK054137.1 | - | 9 - 10 | 1442 | 139 | ||
| BC109135.2 | - | 9 - 10 | 1278 | 40039321 | 139 | |
| M20658.1 | - | 9 - 10 | 1372 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1r1
Genomic Map